Assay Details
Gene Symbol: YPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028827940.1 | XP_028683773.1 | 501 | 89 | ||
GenBank mRNA | JU320893.1 | - | 2 - 3 | 445 | 89 | |
JU471054.1 | - | 2 - 3 | 393 | 89 | ||
JV703771.1 | - | 2 - 3 | 452 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
