Assay Details
Gene Symbol: HRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152901.2 | XP_015008387.1 | 329 | 61 | ||
GenBank mRNA | JV524284.1 | - | 1 - 2 | 65 | 61 | |
JV597153.1 | - | 1 - 2 | 105 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
