Assay Details
Gene Symbol: H6PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147201.2 | XP_015002687.1 | 283 | 75 | ||
GenBank mRNA | JV076197.1 | - | 1 - 2 | 264 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015147201.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
