Assay Details
Gene Symbol: H6PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099298.4 | XP_001099298.2 | 1888 | 116 | ||
XM_015147201.2 | XP_015002687.1 | 1309 | 116 | |||
GenBank mRNA | JV076197.1 | - | 4 - 5 | 1290 | 116 | |
JV666219.1 | - | 4 - 5 | 1262 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
