Assay Details
Gene Symbol: VKORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126009.2 | XP_014981495.2 | 533 | 150 | ||
GenBank mRNA | JU477156.1 | - | 2 - 3 | 518 | 150 | |
JV045678.1 | - | 2 - 3 | 392 | 150 | ||
JV593256.1 | - | 2 - 3 | 468 | 150 | ||
JV661119.1 | - | 2 - 3 | 520 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
