Assay Details
Gene Symbol: LOC703700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145488.2 | XP_015000974.2 | 414 | 120 | ||
GenBank mRNA | JV647133.1 | - | 1 - 2 | 47 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015145488.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
