Assay Details
Gene Symbol: LOC703700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145482.2 | XP_015000968.2 | 1572 | 107 | ||
XM_015145483.2 | XP_015000969.2 | 1595 | 107 | |||
XM_015145485.2 | XP_015000971.2 | 353 | 107 | |||
XM_015145486.2 | XP_015000972.2 | 472 | 107 | |||
XM_015145487.2 | XP_015000973.2 | 448 | 107 | |||
XM_015145488.2 | XP_015000974.2 | 630 | 107 | |||
XM_015145489.2 | XP_015000975.2 | 2022 | 107 | |||
XM_028852705.1 | XP_028708538.1 | 1121 | 107 | |||
XM_028852706.1 | XP_028708539.1 | 1087 | 107 | |||
XM_028852707.1 | XP_028708540.1 | 1893 | 107 | |||
XM_028852708.1 | XP_028708541.1 | 2021 | 107 | |||
XM_028852709.1 | XP_028708542.1 | 2022 | 107 | |||
XM_028852710.1 | XP_028708543.1 | 332 | 107 | |||
GenBank mRNA | JU320776.1 | - | 2 - 3 | 262 | 107 | |
JU320777.1 | - | 2 - 3 | 262 | 107 | ||
JV647133.1 | - | 2 - 3 | 263 | 107 | ||
JV711047.1 | - | 2 - 3 | 284 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
