Assay Details
Gene Symbol: EBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015121087.2 | XP_014976573.2 | 395 | 100 | ||
GenBank mRNA | JV484637.1 | - | 2 - 3 | 132 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
