Assay Details
Gene Symbol: GAREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111866.2 | XP_014967352.2 | 287 | 96 | ||
GenBank mRNA | JV674109.1 | - | 217 | 96 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GAREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111866.2 | XP_014967352.2 | 287 | 96 | ||
GenBank mRNA | JV674109.1 | - | 217 | 96 |