Assay Details
Gene Symbol: LHFPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136222.2 | XP_014991708.1 | 67 | |||
XM_028847635.1 | XP_028703468.1 | 67 | ||||
GenBank mRNA | JV574705.1 | - | 217 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015136222.2, XM_028847635.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
