Assay Details
Gene Symbol: ALKAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832030.1 | XP_028687863.1 | 488 | 96 | ||
XM_028832031.1 | XP_028687864.1 | 488 | 96 | |||
GenBank mRNA | JV058622.1 | - | 139 | 96 | ||
JV648964.1 | - | 261 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
