Assay Details
Gene Symbol: OGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148051.2 | XP_015003537.1 | 876 | 66 | ||
XM_015148052.2 | XP_015003538.1 | 876 | 66 | |||
XM_028826799.1 | XP_028682632.1 | 876 | 66 | |||
GenBank mRNA | JU320291.1 | - | 1 - 2 | 604 | 66 | |
JU320292.1 | - | 1 - 2 | 604 | 66 | ||
JU472439.1 | - | 1 - 2 | 548 | 66 | ||
JV043778.1 | - | 1 - 2 | 609 | 66 | ||
JV705461.1 | - | 1 - 2 | 335 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
