Assay Details
Gene Symbol: ECM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117264.2 | XP_014972750.2 | 1805 | 93 | ||
XM_015117265.2 | XP_014972751.2 | 1774 | 93 | |||
XM_015117266.2 | XP_014972752.2 | 1739 | 93 | |||
XM_015117267.2 | XP_014972753.2 | 1682 | 93 | |||
GenBank mRNA | JU326114.1 | - | 742 | 93 | ||
JV053796.1 | - | 1145 | 93 | |||
JV644343.1 | - | 1665 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
