Assay Details
Gene Symbol: MYL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149055.2 | XP_015004541.1 | 144 | 87 | ||
GenBank mRNA | JU332496.1 | - | 1 - 2 | 59 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015149055.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
