Assay Details
Gene Symbol: MSANTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137898.2 | XP_014993384.2 | 479 | 116 | ||
GenBank mRNA | JV580553.1 | - | 296 | 116 | ||
JV615391.1 | - | 412 | 116 |

More Information
Set Membership: |
![]() ![]() |
