Assay Details
Gene Symbol: MRTFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150425.2 | XP_015005911.2 | 556 | 79 | ||
XM_015150426.2 | XP_015005912.2 | 636 | 79 | |||
XM_015150427.2 | XP_015005913.2 | 574 | 79 | |||
XM_028827670.1 | XP_028683503.1 | 542 | 79 | |||
GenBank mRNA | JU476551.1 | - | 4 - 5 | 520 | 79 | |
JV047766.1 | - | 4 - 5 | 447 | 79 | ||
JV667432.1 | - | 4 - 5 | 602 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015150426.2, XM_015150427.2, XM_028827670.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
