Assay Details
Gene Symbol: MRTFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150425.2 | XP_015005911.2 | 695 | 84 | ||
XM_015150426.2 | XP_015005912.2 | 775 | 84 | |||
XM_015150427.2 | XP_015005913.2 | 713 | 84 | |||
XM_028827670.1 | XP_028683503.1 | 681 | 84 | |||
GenBank mRNA | JU476551.1 | - | 6 - 7 | 659 | 84 | |
JV047766.1 | - | 6 - 7 | 586 | 84 | ||
JV667432.1 | - | 6 - 7 | 741 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
