Assay Details
Gene Symbol: MRTFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150425.2 | XP_015005911.2 | 1023 | 87 | ||
XM_015150426.2 | XP_015005912.2 | 1103 | 87 | |||
XM_015150427.2 | XP_015005913.2 | 1041 | 87 | |||
XM_028827670.1 | XP_028683503.1 | 1009 | 87 | |||
GenBank mRNA | JU476551.1 | - | 8 - 9 | 987 | 87 | |
JV047766.1 | - | 8 - 9 | 914 | 87 | ||
JV667432.1 | - | 8 - 9 | 1069 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
