Assay Details
Gene Symbol: C1QTNF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139805.2 | XP_014995291.2 | 555 | 76 | ||
XM_028841618.1 | XP_028697451.1 | 555 | 76 | |||
XM_028841619.1 | XP_028697452.1 | 640 | 76 | |||
GenBank mRNA | JV651026.1 | - | 1 - 2 | 95 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
