Assay Details
Gene Symbol: CCNJL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084076.4 | XP_001084076.3 | 1022 | 81 | ||
XM_028849883.1 | XP_028705716.1 | 946 | 81 | |||
XM_028849884.1 | XP_028705717.1 | 904 | 81 | |||
XM_028849885.1 | XP_028705718.1 | 1123 | 81 | |||
XM_028849886.1 | XP_028705719.1 | 1118 | 81 | |||
GenBank mRNA | JV071259.1 | - | 2 - 3 | 457 | 81 | |
JV661391.1 | - | 2 - 3 | 471 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
