Assay Details
Gene Symbol: PWWP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141340.2 | XP_014996826.1 | 1743 | 82 | ||
XM_028849881.1 | XP_028705714.1 | 1794 | 82 | |||
GenBank mRNA | JU326417.1 | - | 3 - 4 | 1362 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
