Assay Details
Gene Symbol: PIFO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148180.2 | XP_015003666.1 | 378 | 63 | ||
GenBank mRNA | JU332916.1 | - | 50 | 63 | ||
JV642241.1 | - | 79 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015148180.2 transcripts.
Set Membership: |
![]() ![]() |
