Assay Details
Gene Symbol: FILIP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015129457.2 | XP_014984943.2 | 266 | 124 | ||
GenBank mRNA | JV049033.1 | - | 1 - 2 | 75 | 124 | |
JV639099.1 | - | 1 - 2 | 62 | 124 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015129457.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
