Assay Details
Gene Symbol: DYDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002805661.3 | XP_002805707.3 | 496 | 95 | ||
XM_015147137.2 | XP_015002623.1 | 451 | 95 | |||
XM_015147138.2 | XP_015002624.1 | 381 | 95 | |||
XM_028826413.1 | XP_028682246.1 | 422 | 95 | |||
XM_028826414.1 | XP_028682247.1 | 476 | 95 | |||
XM_028826415.1 | XP_028682248.1 | 402 | 95 | |||
GenBank mRNA | JV725065.1 | - | 356 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
