Assay Details
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015131253.2 | XP_014986739.1 | 308 | 108 | ||
GenBank mRNA | JU334821.1 | - | 212 | 108 | ||
JU475849.1 | - | 208 | 108 | |||
JV047265.1 | - | 210 | 108 | |||
JV583679.1 | - | 168 | 108 | |||
JV653091.1 | - | 217 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
