Assay Details
Gene Symbol: PPM1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001116776.4 | XP_001116776.1 | 6232 | 87 | ||
XM_028829688.1 | XP_028685521.1 | 5800 | 87 | |||
GenBank mRNA | JV552023.1 | - | 1 | 3616 | 87 | |
JV623231.1 | - | 1 | 1923 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
