Assay Details
Gene Symbol: CDIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141815.2 | XP_014997301.1 | 534 | 104 | ||
XM_015141816.2 | XP_014997302.1 | 534 | 104 | |||
XM_015141817.2 | XP_014997303.1 | 534 | 104 | |||
XM_015141818.2 | XP_014997304.2 | 255 | 104 | |||
XM_015141819.2 | XP_014997305.1 | 534 | 104 | |||
XM_015141820.2 | XP_014997306.1 | 488 | 104 | |||
XM_015141822.2 | XP_014997308.1 | 534 | 104 | |||
XM_028850721.1 | XP_028706554.1 | 269 | 104 | |||
GenBank mRNA | JU321014.1 | - | 515 | 104 | ||
JU321187.1 | - | 515 | 104 | |||
JU321192.1 | - | 515 | 104 | |||
JU477516.1 | - | 405 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015141818.2, XM_015141820.2, XM_028850721.1 transcripts.
Set Membership: |
![]() ![]() |
