Assay Details
Gene Symbol: CDIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141815.2 | XP_014997301.1 | 612 | 75 | ||
XM_015141816.2 | XP_014997302.1 | 612 | 75 | |||
XM_015141817.2 | XP_014997303.1 | 612 | 75 | |||
XM_015141818.2 | XP_014997304.2 | 333 | 75 | |||
XM_015141819.2 | XP_014997305.1 | 612 | 75 | |||
XM_015141820.2 | XP_014997306.1 | 566 | 75 | |||
XM_015141822.2 | XP_014997308.1 | 612 | 75 | |||
XM_028850721.1 | XP_028706554.1 | 347 | 75 | |||
XM_028850722.1 | XP_028706555.1 | 571 | 75 | |||
GenBank mRNA | JU321014.1 | - | 5 - 6 | 593 | 75 | |
JU321187.1 | - | 5 - 6 | 593 | 75 | ||
JU321192.1 | - | 5 - 6 | 593 | 75 | ||
JU477516.1 | - | 5 - 6 | 483 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
