Assay Details
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028837492.1 | XP_028693325.1 | 99 | 102 | ||
XM_028837495.1 | XP_028693328.1 | 113 | 102 | |||
XM_028837498.1 | XP_028693331.1 | 99 | 102 | |||
GenBank mRNA | JU323153.1 | - | 62 | 102 | ||
JV054780.1 | - | 57 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028837492.1, XM_028837495.1, XM_028837498.1 transcripts.
Set Membership: |
![]() ![]() |
