Assay Details
Gene Symbol: EEF1AKMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028837495.1 | XP_028693328.1 | 645 | 79 | ||
GenBank mRNA | JU323153.1 | - | 594 | 79 | ||
JU473625.1 | - | 571 | 79 | |||
JV054780.1 | - | 589 | 79 | |||
JV667218.1 | - | 564 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
