Assay Details
Gene Symbol: BCL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028830020.1 | XP_028685853.1 | 1146 | 133 | ||
XM_028830021.1 | XP_028685854.1 | 459 | 133 | |||
GenBank mRNA | JU321668.1 | - | 177 | 133 | ||
JU472189.1 | - | 191 | 133 | |||
JU472190.1 | - | 191 | 133 | |||
JV589890.1 | - | 246 | 133 |

More Information
Set Membership: |
![]() ![]() |
