Assay Details
Gene Symbol: BCL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028830021.1 | XP_028685854.1 | 928 | 82 | ||
GenBank mRNA | JU321668.1 | - | 646 | 82 | ||
JU472190.1 | - | 660 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
