Assay Details
Gene Symbol: SLC46A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015117058.2 | XP_014972544.2 | 1586 | 102 | ||
GenBank mRNA | JU472351.1 | - | 1585 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
