Assay Details
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028846586.1 | XP_028702419.1 | 6109 | 71 | ||
GenBank mRNA | JU480552.1 | - | 226 | 71 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028846586.1 | XP_028702419.1 | 6109 | 71 | ||
GenBank mRNA | JU480552.1 | - | 226 | 71 |