Assay Details
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028846586.1 | XP_028702419.1 | 6383 | 154 | ||
GenBank mRNA | JU480552.1 | - | 500 | 154 |

More Information
Set Membership: |
![]() ![]() |
