Assay Details
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123836.2 | XP_014979322.1 | 1158 | 75 | ||
XM_015123837.2 | XP_014979323.1 | 1173 | 75 | |||
XM_015123838.2 | XP_014979324.1 | 1150 | 75 | |||
XM_015123839.2 | XP_014979325.1 | 1102 | 75 | |||
XM_015123840.2 | XP_014979326.1 | 1078 | 75 | |||
XM_015123841.2 | XP_014979327.1 | 1142 | 75 | |||
XM_015123842.2 | XP_014979328.1 | 1349 | 75 | |||
XM_028839582.1 | XP_028695415.1 | 1043 | 75 | |||
GenBank mRNA | JV597804.1 | - | 112 | 75 | ||
JV632883.1 | - | 124 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
