Assay Details
Gene Symbol: LPAR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150911.2 | XP_015006397.2 | 2261 | 113 | ||
XM_028829203.1 | XP_028685036.1 | 2428 | 113 | |||
GenBank mRNA | JV719240.1 | - | 1 | 718 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
