Assay Details
Gene Symbol: HCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839480.1 | XP_028695313.1 | 1883 | 121 | ||
GenBank mRNA | JU346815.1 | - | 1395 | 121 | ||
JV589999.1 | - | 1418 | 121 | |||
JV692570.1 | - | 1421 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
