Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015150288.2 | XP_015005774.2 | 1569 | 94 | ||
XM_015150290.2 | XP_015005776.2 | 1093 | 94 | |||
XM_028827740.1 | XP_028683573.1 | 593 | 94 | |||
XM_028827741.1 | XP_028683574.1 | 735 | 94 | |||
XM_028827742.1 | XP_028683575.1 | 529 | 94 | |||
GenBank mRNA | JU323622.1 | - | 421 | 94 | ||
JV641056.1 | - | 503 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
