Assay Details
Gene Symbol: P2RY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115162.2 | XP_014970648.1 | 2201 | 70 | ||
XM_015115165.2 | XP_014970651.1 | 1864 | 70 | |||
XM_015115167.2 | XP_014970653.1 | 693 | 70 | |||
XM_015115168.2 | XP_014970654.1 | 669 | 70 | |||
XM_015115169.2 | XP_014970655.1 | 531 | 70 | |||
XM_015115170.2 | XP_014970656.1 | 2030 | 70 | |||
XM_015115171.2 | XP_014970657.1 | 593 | 70 | |||
XM_028833803.1 | XP_028689636.1 | 1729 | 70 | |||
XM_028833804.1 | XP_028689637.1 | 2332 | 70 | |||
XM_028833805.1 | XP_028689638.1 | 655 | 70 | |||
GenBank mRNA | JU324139.1 | - | 3 | 418 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
