Assay Details
Gene Symbol: KRBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108465.4 | XP_001108465.2 | 433 | 100 | ||
XM_015131215.2 | XP_014986701.1 | 436 | 100 | |||
XM_015131216.2 | XP_014986702.1 | 346 | 100 | |||
XM_015131217.2 | XP_014986703.1 | 445 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
