Assay Details
Gene Symbol: LOC695594 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002803500.3 | XP_002803546.1 | 504 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002803500.3 transcripts.
Set Membership: |
![]() ![]() |
