Assay Details
Gene Symbol: LIPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082459.4 | XP_001082459.3 | 989 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
