Assay Details
Gene Symbol: TMBIM7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134232.2 | XP_014989718.1 | 363 | 102 | ||
XM_015134233.2 | XP_014989719.1 | 358 | 102 | |||
XM_015134234.2 | XP_014989720.1 | 353 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015134232.2, XM_015134233.2, XM_015134234.2 transcripts.
Set Membership: |
![]() ![]() |
