Assay Details
Gene Symbol: TMBIM7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134232.2 | XP_014989718.1 | 866 | 65 | ||
XM_015134233.2 | XP_014989719.1 | 861 | 65 |

More Information
Set Membership: |
![]() ![]() |
