Assay Details
Gene Symbol: TMBIM7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134232.2 | XP_014989718.1 | 1134 | 65 | ||
XM_015134234.2 | XP_014989720.1 | 998 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
