Assay Details
Gene Symbol: IL37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832311.1 | XP_028688144.1 | 846 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028832311.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
