Assay Details
Gene Symbol: LOC713290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109975.4 | XP_001109975.1 | 100 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001109975.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
