Assay Details
Gene Symbol: LOC100425941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028831320.1 | XP_028687153.1 | 321 | 108 | ||
XM_028831321.1 | XP_028687154.1 | 325 | 108 | |||
XM_028831322.1 | XP_028687155.1 | 323 | 108 | |||
XM_028831323.1 | XP_028687156.1 | 327 | 108 |

More Information
Set Membership: |
![]() ![]() |
