Assay Details
Gene Symbol: CDC42EP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015124869.2 | XP_014980355.2 | 395 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015124869.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
